missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,5-Dichloropentane, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
25 g
100 g
500 g
Description
1,5-Dichloropentane is used in the preparation of symmetrical sulfide, tetrydrothiopyran (thiane) by reaction with sodium sulfide.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
1,5-Dichloropentane is used in the preparation of symmetrical sulfide, tetrydrothiopyran (thiane) by reaction with sodium sulfide.
Solubility
Miscible with ethanol, ether and chloroform. Immiscible with water.
Notes
Incompatible with strong oxidizing agents and strong bases.
1,5-Dichloropentane is used in the preparation of symmetrical sulfide, tetrydrothiopyran (thiane) by reaction with sodium sulfide.
Solubility
Miscible with ethanol, ether and chloroform. Immiscible with water.
Notes
Incompatible with strong oxidizing agents and strong bases.
Specifications
Melting Point | -72°C |
Density | 1.103 |
Boiling Point | 182°C |
Flash Point | 26°C (78°F) |
Linear Formula | Cl(CH2)5Cl |
Refractive Index | 1.457 |
Quantity | 500 g |
UN Number | UN1152 |
Beilstein | 1732340 |
Solubility Information | Miscible with ethanol,ether and chloroform. Immiscible with water. |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'