missing translation for 'onlineSavingsMsg'
Learn More

1,5-Dichlorohexamethyltrisiloxane (~90%), TRC

Product Code. 30382856
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30382856 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30382856 Supplier Toronto Research Chemicals Supplier No. TRCD43611850MG

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 1,5-Dichlorohexamethyltrisiloxane
CAS 3582-71-6
Molecular Formula C6H18Cl2O2Si3
Quantity 50 mg
Synonym 1,5-Dichloro-1,1,3,3,5,5-hexamethyltrisiloxane, 1,1,3,3,5,5-Hexamethyl-1,5-dichlorotrisiloxane, 1-Chloro-3-chlorodimethylsilyloxy-1,1,3,3-tetramethyldisiloxane, Bis(chlorodimethylsiloxy)dimethylsilane, Hexamethyl-1,5-dichlorotrisiloxane
InChI Formula InChI=1S/C6H18Cl2O2Si3/c1-11(2,7)9-13(5,6)10-12(3,4)8/h1-6H3
SMILES C[Si](C)(Cl)O[Si](C)(C)O[Si](C)(C)Cl
IUPAC Name bis[[chloro(dimethyl)silyl]oxy]-dimethylsilane
Molecular Weight (g/mol) 277.36
Formula Weight 275.9992
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.