missing translation for 'onlineSavingsMsg'
Learn More

1,5-Dichloro-2,4-dinitrobenzene, 97%

Product Code. 11386936
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11386936 5 g 5g
11396936 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11386936 Supplier Thermo Scientific Alfa Aesar Supplier No. L06652.06
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 3698-83-7 | C6H2Cl2N2O4 | 236.992 g/mol

1,5-Dichloro-2,4-dinitrobenzene is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,5-Dichloro-2,4-dinitrobenzene is used as pharmaceutical intermediate.

Solubility
Insoluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3698-83-7
Molecular Formula C6H2Cl2N2O4
Molecular Weight (g/mol) 236.992
MDL Number MFCD00024186
InChI Key ZPXDNSYFDIHPOJ-UHFFFAOYSA-N
Synonym 1,3-dichloro-4,6-dinitrobenzene, benzene, 1,5-dichloro-2,4-dinitro, 2,4-dichloro-1,5-dinitrobenzene, 4,6-dichloro-1,3-dinitrobenzene, unii-6enc7c8zr0, 6enc7c8zr0, 1,5-dichloro-2,4-dinitro-benzene, 1,3,-dichloro-4,6-dinitrobenzene, 1,5-dichlor-2,4-dinitrobenzol, 2,5-dinitrobenzene
PubChem CID 77286
IUPAC Name 1,5-dichloro-2,4-dinitrobenzene
SMILES C1=C(C(=CC(=C1[N+](=O)[O-])Cl)Cl)[N+](=O)[O-]

Specifications

Melting Point 98°C to 103°C
Quantity 5 g
UN Number UN2811
Beilstein 1685438
Formula Weight 237
Percent Purity 97%
Chemical Name or Material 1,5-Dichloro-2,4-dinitrobenzene

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.