missing translation for 'onlineSavingsMsg'
Learn More

1,5-Dibromopentane, TRC

Product Code. 30388081
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Quantity:
10 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30388081 25 g
30385045 100 g
30387958 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 missing translation for 'options'
This item is not returnable. View return policy
Product Code. 30388081 missing translation for 'mfr' Toronto Research Chemicals missing translation for 'supplierNo' TRCD42559025G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 111-24-0
Molecular Formula C5 H10 Br2
Molecular Weight (g/mol) 229.9409
Synonym Pentane, 1,5-dibromo-, 1,5-Dibromopentane, 1,5-Pentanedibromide, NSC 5373, Pentamethylene bromide, Pentamethylene dibromide
IUPAC Name 1,5-dibromopentane
SMILES BrCCCCCBr

Specifications

Quantity 25 g
InChI Formula InChI=1S/C5H10Br2/c6-4-2-1-3-5-7/h1-5H2
Formula Weight 227.9149
Chemical Name or Material 1,5-Dibromopentane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.