Learn More
1,5-Dibromopentane, 98%
Description
1,5-Dibromopentane is used in the preparation of 1,5-di-Grignard reagent and novel spirocyclic pyrrolidones. It acts as a growth supplement for Yarrowia lipolytica 3589, a tropical marine yeast. It is involved in the synthesis of N-alkylated piperidine as well as tetrydrothiopyran (thiane) by reaction with primary amine and sodium sulfide respectively.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
1,5-Dibromopentane is used in the preparation of 1,5-di-Grignard reagent and novel spirocyclic pyrrolidones. It acts as a growth supplement for Yarrowia lipolytica 3589, a tropical marine yeast. It is involved in the synthesis of N-alkylated piperidine as well as tetrydrothiopyran (thiane) by reaction with primary amine and sodium sulfide respectively.
Solubility
Miscible with water, slightly miscible with chloroform and benzene.
Notes
Incompatible with strong oxidizing agents and strong bases.
Specifications
Specifications
| Melting Point | -34°C |
| Density | 1.688 |
| Boiling Point | 224°C |
| Flash Point | 79°C (174°F) |
| Refractive Index | 1.5122 |
| Quantity | 500 g |
| Beilstein | 1209245 |
| Synonym | pentamethylene bromide, pentane, 1,5-dibromo, pentamethylene dibromide, unii-b8q228qys1, pentane-1,5-dibromide, 1,5-dibromo-pentane, 1,5-pentane dibromide, pentane,1,5-dibromo, br ch2 5br, ccris 8918 |
| Solubility Information | Miscible with water,slightly miscible with chloroform and benzene. |
| Formula Weight | 229.95 |
| Show More |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.