missing translation for 'onlineSavingsMsg'
Learn More

1,5-Dibromo-3-methylpentane, 98+%

Product Code. 11428887
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 11428887

Brand: Thermo Scientific Alfa Aesar B20521.14

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 4457-72-1 | C6H12Br2 | 243.97 g/mol

1,5-Dibromo-3-methylpentane is used as a chemical reagent, pharmaceutical intermediate, pharmaceutical research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,5-Dibromo-3-methylpentane is used as a chemical reagent, pharmaceutical intermediate, pharmaceutical research.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents, strong bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4457-72-1
Molecular Formula C6H12Br2
Molecular Weight (g/mol) 243.97
MDL Number MFCD00039200
InChI Key YDPZWUMQKMLLHC-UHFFFAOYSA-N
Synonym 3-methyl-1,5-dibromopentane, 1,5-dibromo-3-methyl-pentane, pentane, 1,5-dibromo-3-methyl, acmc-20ap1m, pentane,1,5-dibromo-3-methyl, 1,5-dibromo-3-methylpentane 5g
PubChem CID 138258
IUPAC Name 1,5-dibromo-3-methylpentane
SMILES CC(CCBr)CCBr

Specifications

Density 1.58
Boiling Point 233°C
Flash Point 101°C (214°F)
Linear Formula BrCH2CH2CH(CH3)CH2CH2Br
Refractive Index 1.508
Quantity 25 g
Solubility Information Insoluble in water.
Formula Weight 243.98
Percent Purity ≥98%
Chemical Name or Material 1,5-Dibromo-3-methylpentane
compliance-icons
Product Identifier
  • 1,5-Dibromo-3-methylpentane
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
  • P362+P364-Take off contaminated clothing and wash it before reuse.
  • P403-Store in a well-ventilated place.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : 1,5-Dibromo-3-methylpentane

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.