missing translation for 'onlineSavingsMsg'
Learn More

1,5-Dibromo-1-(2-fluorophenyl)-2-pentanone, TRC

Product Code. 30386451
Change view
Click to view available options
Quantity:
10 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30386451 100 mg
30375591 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30386451 Supplier Toronto Research Chemicals Supplier No. TRCD425575100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1373350-57-2
Molecular Formula C11H11Br2FO
Molecular Weight (g/mol) 338.01
Synonym Prasugrel Impurity
IUPAC Name 1,5-dibromo-1-(2-fluorophenyl)pentan-2-one
SMILES FC1=CC=CC=C1C(Br)C(CCCBr)=O

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C11H11Br2FO/c12-7-3-6-10(15)11(13)8-4-1-2-5-9(8)14/h1-2,4-5,11H,3,6-7H2
Formula Weight 335.92
Chemical Name or Material 1,5-Dibromo-1-(2-fluorophenyl)-2-pentanone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.