missing translation for 'onlineSavingsMsg'
Learn More

1,5-Diaminoanthraquinone, 90+%

Product Code. 11453108
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11453108 25 g 25g
11463108 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11453108 Supplier Thermo Scientific Alfa Aesar Supplier No. A18840.14
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 129-44-2 | C14H10N2O2 | 238.246 g/mol

1,5-Diaminoanthraquinone (DAAQ) may be used in the preparation of vertical nanowire arrays. It may be used in the synthesis of poly(1,5-diaminoanthraquinone) (PDAA) particles, via chemically oxidative polymerization of DAAQ.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,5-Diaminoanthraquinone (DAAQ) may be used in the preparation of vertical nanowire arrays. It may be used in the synthesis of poly(1,5-diaminoanthraquinone) (PDAA) particles, via chemically oxidative polymerization of DAAQ.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 129-44-2
Molecular Formula C14H10N2O2
Molecular Weight (g/mol) 238.246
MDL Number MFCD00001226
InChI Key VWBVCOPVKXNMMZ-UHFFFAOYSA-N
Synonym 1,5-diaminoanthraquinone, smoke red f, 1,5-diaminoanthrachinon, c.i. disperse red ii, 1,5-anthraquinonyldiamine, 9,10-anthracenedione, 1,5-diamino, 1,5-diamino-9,10-anthraquinone, 1,5-daa, 1,5-daa russian, unii-3zxx3hk358
PubChem CID 8513
IUPAC Name 1,5-diaminoanthracene-9,10-dione
SMILES C1=CC2=C(C(=C1)N)C(=O)C3=C(C2=O)C(=CC=C3)N

Specifications

Melting Point ∼320°C (decomposition)
Odor Odorless
Quantity 25 g
Solubility Information Insoluble in water.
Formula Weight 238.25
Percent Purity 97%
Chemical Name or Material 1,5-Diaminoanthraquinone

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.