missing translation for 'onlineSavingsMsg'
Learn More

1,5-Diamino-4,8-dihydroxy-9,10-dihydroanthracene-9,10-dione, Tech., Thermo Scientific™

Product Code. 10213662
Change view
Click to view available options
Quantity:
10 g
50 g
100 g
Unit Size:
100g
10g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10213662 10 g 10g
10131082 50 g 50g
10554695 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10213662 Supplier Thermo Scientific Maybridge Supplier No. SB01090EA
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 145-49-3
Molecular Formula C14H10N2O4
Molecular Weight (g/mol) 270.244
MDL Number MFCD00013722
InChI Key HSYLKWSCFRLSKB-UHFFFAOYSA-N
Synonym 1,5-diamino-4,8-dihydroxyanthraquinone, diaminoanthrarufin, 1,5-diaminoanthrarufin, 4,8-diaminoanthrarufin, 1,5-dihydroxy-4,8-diaminoanthraquinone, anthrarufin, 4,8-diamino, unii-q7yvg56q18, ccris 5599, 4,8-diamino-1,5-dihydroxyanthraquinone, 1,5-diamino-4,8-dihydroxy-anthracene-9,10-dione
PubChem CID 67351
IUPAC Name 1,5-diamino-4,8-dihydroxyanthracene-9,10-dione
SMILES C1=CC(=C2C(=C1N)C(=O)C3=C(C=CC(=C3C2=O)N)O)O

Specifications

Packaging Amber Glass Bottle
Quantity 10 g
Formula Weight 270.24
Percent Purity Tech.
Grade Technical
Chemical Name or Material 1,5-Diamino-4,8-dihydroxy-9,10-dihydroanthracene-9,10-dione
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.