missing translation for 'onlineSavingsMsg'
Learn More

1,5-Cyclooctadienebis(methyldiphenylphosphine)iridium(I) hexafluorophosphate

Product Code. 11373477
Change view
Click to view available options
Quantity:
0.5 g
2 g
Unit Size:
0.5g
2g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11373477 2 g 2g
11383477 0.5 g 0.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11373477 Supplier Thermo Scientific Alfa Aesar Supplier No. 018809.04
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Hydrogenation | CAS: 38465-86-0 | C34H38F6IrP3 | 845.81 g/mol

1,5-Cyclooctadienebis(methyldiphenylphosphine)iridium(I) hexafluorophosphate is used as a catalyst for isomerization of allylic ethers of glycerol and glucose to the corresponding 1-propenyl ethers. It is also used as a catalyst in hydrogenation reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 38465-86-0
Molecular Formula C34H38F6IrP3
Molecular Weight (g/mol) 845.81
MDL Number MFCD00064800
InChI Key LXKHQEQLBSJJCO-JXNOXZOESA-N
Synonym 1,5-cyclooctadiene bis methyldiphenylphosphine iridium i hexafluorophosphate, 1,5-cyclooctadiene bis methyldiphenylphosphine ir, 1,5-cyclooctadienebis methyldiphenylphosphine iridium i hexafluorophosphate, 1z,5z-cycloocta-1,5-diene; iridium; methyl diphenyl phosphane; hexafluorophosphate, iridium 1+ 1,5-cyclooctadiene, z,z-bis methyldiphenylphosphane hexafluorophosphate
PubChem CID 5702663
IUPAC Name (1Z,5Z)-cycloocta-1,5-diene;iridium;methyl(diphenyl)phosphane;hexafluorophosphate
SMILES [Ir+].F[P-](F)(F)(F)(F)F.C1C\C=C/CC\C=C/1.CP(C1=CC=CC=C1)C1=CC=CC=C1.CP(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 224°C (decomposition)
Quantity 2 g
Sensitivity Air and moisture sensitive
Solubility Information Soluble in acetone; Insoluble in water.
Formula Weight 845.79
Physical Form Crystalline
Chemical Name or Material 1,5-Cyclooctadienebis(methyldiphenylphosphine)iridium(I) hexafluorophosphate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.