missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,5-Cyclooctadiene, TRC
Click to view available options
Quantity:
50 g
Unit Size:
50g
Specifications
Specifications
| Chemical Name or Material | 1,5-Cyclooctadiene |
| CAS | 111-78-4 |
| Molecular Formula | C8H12 |
| Quantity | 50 g |
| Synonym | 1,5-COD, cis-1,5-Cyclooctadiene, COD |
| InChI Formula | InChI=1S/C8H12/c1-2-4-6-8-7-5-3-1/h1-2,7-8H,3-6H2 |
| SMILES | C1CC=CCCC=C1 |
| IUPAC Name | cycloocta-1,5-diene |
| Molecular Weight (g/mol) | 108.18 |
| Formula Weight | 108.0939 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction