missing translation for 'onlineSavingsMsg'
Learn More

(1,5-Cyclooctadiene)(hexafluoroacetylacetonato)iridium(I), TRC

Product Code. 30368151
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30368151 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30368151 Supplier Toronto Research Chemicals Supplier No. TRCC98458810MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material (1,5-Cyclooctadiene)(hexafluoroacetylacetonato)iridium(I)
CAS 34801-95-1
Molecular Formula C8 H12 . C7 H9 F6 Ir O2
Quantity 10 mg
InChI Formula InChI=1S/C8H12.C5H2F6O2.2CH4.Ir/c1-2-4-6-8-7-5-3-1;6-4(7,8)2(12)1-3(13)5(9,10)11;;;/h1-2,7-8H,3-6H2;1,12H;2*1H4;/q;;;;+1/p-1/b2-1-,8-7-;;;;
SMILES FC(F)(F)C1=[O][Ir](=[O]C(=C1)C(F)(F)F)([CH4])[CH4].C2C\C=C/CC\C=C/2
IUPAC Name (1Z,5Z)-cycloocta-1,5-diene;2,2-di(methyl)-4,6-bis(trifluoromethyl)-1λ3,3λ3-dioxa-2λ5-iridacyclohexa-1,3,5-triene
Molecular Weight (g/mol) 539.534
Formula Weight 540.107
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.