missing translation for 'onlineSavingsMsg'
Learn More

1-(5-Chloro-2-pyridyl)piperazine, 99%

Product Code. 11454900
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
This item is not returnable. View return policy

Product Code. 11454900

Brand: Thermo Scientific Alfa Aesar H50862.MD

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 87394-65-8 | C9H12ClN3 | 197.666 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 87394-65-8
Molecular Formula C9H12ClN3
Molecular Weight (g/mol) 197.666
MDL Number MFCD04115049
InChI Key ZTLOZFLZBKZABP-UHFFFAOYSA-N
Synonym 1-5-chloropyridin-2-yl piperazine, 1-5-chloro-2-pyridyl piperazine, 1-5-chloro-pyridin-2-yl-piperazine, 5-chloro-2-piperazin-1-yl pyridine, 1-5-chloro-2-pyridinyl piperazine, acmc-20ap9z, 2-piperazino 5-chloro pyridine, 5-chloro-2-pyridyl piperazine, 5-chloro-2-1-piperazinyl pyridine, 1-5-chloro-2-pyridinyl-piperazine
PubChem CID 3805301
IUPAC Name 1-(5-chloropyridin-2-yl)piperazine
SMILES C1CN(CCN1)C2=NC=C(C=C2)Cl

Specifications

Quantity 250 mg
Formula Weight 197.67
Percent Purity 99%
Chemical Name or Material 1-(5-Chloro-2-pyridyl)piperazine

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.