missing translation for 'onlineSavingsMsg'
Learn More

1,5,8,12-Tetrakis[4,6-bis(N-butyl-N-1,2,2,6,6-pentamethyl-4-piperidylamino)-1,3,5-triazin-2-yl]-1,5,8,12-tetraazadodecane, TRC

Product Code. 30137525
Change view
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30137525 10 g 10g
30004700 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30137525 Supplier Toronto Research Chemicals Supplier No. TRCT29641010G
 Restricted Item
Estimated Shipment: 13-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 106990-43-6
Molecular Formula C132 H250 N32
Molecular Weight (g/mol) 2285.61
Synonym 1,3,5-Triazine-2,4,6-triamine, N2,N2'-1,2-ethanediylbis[N2-[3-[[4,6-bis[butyl(1,2,2,6,6-pentamethyl-4-piperidinyl)amino]-1,3,5-triazin-2-yl]amino]propyl]-N4,N6-dibutyl-N4,N6-bis(1,2,2,6,6-pentamethyl-4-piperidinyl)- (ACI), 1,3,5-Triazine-2,4,6-triamine, N,N' ' '-1,2-ethanediylbis[N-[3-[[4,6-bis[butyl(1,2,2,6,6-pentamethyl-4-piperidinyl)amino]-1,3,5-triazin-2-yl]amino]propyl]-N',N' '-dibutyl-N',N' '-bis(1,2,2,6,6-pentamethyl-4-piperidinyl)- (9CI), 1,5,8,12-Tetrakis[4,6-bis(N-butyl-N-1,2,2,6,6-pentamethyl-4-piperidylamino)-1,3,5-triazin-2-yl]-1,5,8,12-tetraazadodecane, 119FL, Antioxidant 119, Antioxidant 228, Chiguard 228, Chimassorb 119, Chimassorb 119FL, Chimassorb NOR 119, Chisorb 519, GW 119, HALS 11, HALS 119, Light Stabilizer 119, Lowilite 19, N,N'N' 'N' ' '-Tetrakis[4,6-bis[butyl(N-methyl-2,2,6,6-tetramethylpiperidin-4-yl)amino]triazin-2-yl]-4,7-diazadecane-1,10-diamine, Sabostab 119FL, Sabostab 119UV, Sabostab UV 119, Tinuvin 119, UV 119
IUPAC Name 6-N-[3-[[4,6-bis[butyl-(1,2,2,6,6-pentamethylpiperidin-4-yl)amino]-1,3,5-triazin-2-yl]-[2-[[4,6-bis[butyl-(1,2,2,6,6-pentamethylpiperidin-4-yl)amino]-1,3,5-triazin-2-yl]-[3-[[4,6-bis[butyl-(1,2,2,6,6-pentamethylpiperidin-4-yl)amino]-1,3,5-triazin-2-yl]amino]propyl]amino]ethyl]amino]propyl]-2-N,4-N-dibutyl-2-N,4-N-bis(1,2,2,6,6-pentamethylpiperidin-4-yl)-1,3,5-triazine-2,4,6-triamine
SMILES CCCCN(C1CC(C)(C)N(C)C(C)(C)C1)c2nc(NCCCN(CCN(CCCNc3nc(nc(n3)N(CCCC)C4CC(C)(C)N(C)C(C)(C)C4)N(CCCC)C5CC(C)(C)N(C)C(C)(C)C5)c6nc(nc(n6)N(CCCC)C7CC(C)(C)N(C)C(C)(C)C7)N(CCCC)C8CC(C)(C)N(C)C(C)(C)C8)c9nc(nc(n9)N(CCCC)C%10CC(C)(C)N(C)C(C)(C)C%10)N(CCCC)C%11CC(C)(C)N(C)C(C)(C)C%11)nc(n2)N(CCCC)C%12CC(C)(C)N(C)C(C)(C)C%12

Specifications

Quantity 10 g
InChI Formula InChI=1S/C132H250N32/c1-49-57-71-157(97-81-117(9,10)147(41)118(11,12)82-97)109-135-105(136-110(143-109)158(72-58-50-2)98-83-119(13,14)148(42)120(15,16)84-98)133-67-65-69-155(107-139-113(161(75-61-53-5)101-89-125(25,26)151(45)126(27,28)90-101)145-114(140-107)162(76-62-54-6)102-91-127(29,30)152(46)128(31,32)92-102)79-80-156(108-141-115(163(77-63-55-7)103-93-129(33,34)153(47)130(35,36)94-103)146-116(142-108)164(78-64-56-8)104-95-131(37,38)154(48)132(39,40)96-104)70-66-68-134-106-137-111(159(73-59-51-3)99-85-121(17,18)149(43)122(19,20)86-99)144-112(138-106)160(74-60-52-4)100-87-123(21,22)150(44)124(23,24)88-100/h97-104H,49-96H2,1-48H3,(H,133,135,136,143)(H,134,137,138,144)
Formula Weight 2284.0546
Chemical Name or Material 1,5,8,12-Tetrakis[4,6-bis(N-butyl-N-1,2,2,6,6-pentamethyl-4-piperidylamino)-1,3,5-triazin-2-yl]-1,5,8,12-tetraazadodecane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.