missing translation for 'onlineSavingsMsg'
Learn More

1-[(4R,5R)-4,5-Dihydroxy-L-ornithine]-Echinocandin B Hydrochloride, TRC

Product Code. 30386599
Change view
Click to view available options
Quantity:
50 mg
100 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30386599 100 mg
30383757 50 mg
30375627 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30386599 Supplier Toronto Research Chemicals Supplier No. TRCE325645100MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1029890-89-8
Molecular Formula C34H51N7O15 . HCl
Molecular Weight (g/mol) 834.3
Synonym Anidulafungin Nucleus
SMILES Cl.C[C@@H](O)[C@@H]1NC(=O)C(NC(=O)[C@@H]2C[C@@H](O)CN2C(=O)C(NC(=O)[C@@H](N)C[C@@H](O)[C@@H](O)NC(=O)[C@@H]3[C@@H](O)[C@@H](C)CN3C1=O)[C@@H](C)O)[C@H](O)[C@@H](O)c4ccc(O)cc4

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C34H51N7O15.ClH/c1-12-10-41-24(25(12)47)32(54)39-30(52)20(46)9-18(35)28(50)36-21(13(2)42)33(55)40-11-17(45)8-19(40)29(51)38-23(31(53)37-22(14(3)43)34(41)56)27(49)26(48)15-4-6-16(44)7-5-15;/h4-7,12-14,17-27,30,42-49,52H,8-11,35H2,1-3H3,(H,36,50)(H,37,53)(H,38,51)(H,39,54);1H/t12-,13+,14+,17+,18-,19-,20+,21-,22-,23-,24-,25-,26-,27-,30+;/m0./s1
Formula Weight 833.321
Chemical Name or Material 1-[(4R,5R)-4,5-Dihydroxy-L-ornithine]-Echinocandin B Hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.