missing translation for 'onlineSavingsMsg'
Learn More

1-(4-Trifluoromethylphenyl)pentan-1-one, TRC

Product Code. 30451783
Change view
Click to view available options
Quantity:
250 mg
2.5 g
Unit Size:
2.5g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30451783 250 mg 250mg
30455553 2.5 g 2.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30451783 Supplier Toronto Research Chemicals Supplier No. TRCT791420250MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 42916-66-5
Molecular Formula C12H13F3O
Molecular Weight (g/mol) 230.23
Synonym 1-[4-(Trifluoromethyl)phenyl]-1-pentanone, p-(Trifluoromethyl)valerophenone, 1-(4-Trifluoromethylphenyl)pentan-1-one
IUPAC Name 1-(4-(trifluoromethyl)phenyl)pentan-1-one
SMILES O=C(CCCC)C1=CC=C(C(F)(F)F)C=C1

Specifications

Quantity 250 mg
InChI Formula InChI=1S/C12H13F3O/c1-2-3-4-11(16)9-5-7-10(8-6-9)12(13,14)15/h5-8H,2-4H2,1H3
Formula Weight 230.09
Chemical Name or Material 1-(4-Trifluoromethylphenyl)pentan-1-one
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.