missing translation for 'onlineSavingsMsg'
Learn More

1-[4-(Trifluoromethyl)pyrimidin-2-yl]-1,4-diazepane, 95%, Thermo Scientific™

Product Code. 10737275
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Quantity:
1 g
5 g
missing translation for 'unitSize':
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity missing translation for 'unitSize'
10737275 5 g 5g
10651473 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 missing translation for 'options'
This item is not returnable. View return policy
Product Code. 10737275 missing translation for 'mfr' Thermo Scientific Maybridge missing translation for 'supplierNo' AW00488DE
 Restricted Item
Estimated Shipment: 08-02-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 238403-48-0
Molecular Formula C10H13F3N4
Molecular Weight (g/mol) 246.24
MDL Number MFCD00728997
InChI Key XVAZHAVSZFQOOQ-UHFFFAOYSA-N
Synonym 1-4-trifluoromethyl pyrimidin-2-yl-1,4-diazepane, 1-4-trifluoromethyl-2-pyrimidinyl-1,4-diazepane, 1-4-trifluoromethyl pyrimid-2-yl homopiperazine, 1-4-trifluoromethyl pyrimidin-2-yl homopiperazine, 1h-1,4-diazepine,hexahydro-1-4-trifluoromethyl-2-pyrimidinyl, maybridge1_000269, 1-4-trifluoromethyl pyrimidin-2-yl-1,4-diazaperhydroepine
PubChem CID 2782067
IUPAC Name 1-[4-(trifluoromethyl)pyrimidin-2-yl]-1,4-diazepane
SMILES FC(F)(F)C1=NC(=NC=C1)N1CCCNCC1

Specifications

CAS Min % 94.5
CAS Max % 100.0
Assay Percent Range 95%
Quantity 5 g
Formula Weight 246.23
Percent Purity 95%
Chemical Name or Material 1-[4-(Trifluoromethyl)pyrimidin-2-yl]-1, 4-diazepane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.