missing translation for 'onlineSavingsMsg'
Learn More

1-[4-(Trifluoromethyl)-2-pyrimidinyl]-4-piperidinecarboxylic acid, ≥97%, Thermo Scientific™

Product Code. p-3110084
Click to view available options
Quantity:
1 g
10 g
Packaging:
Amber Glass Bottle
Unit Size:
10g
1g
This item is not returnable. View return policy

Product Code. 10640725

Brand: Thermo Scientific Maybridge AW01250DA

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 465514-39-0
Molecular Formula C11H12F3N3O2
Molecular Weight (g/mol) 275.23
MDL Number MFCD02091615
InChI Key QYAIISDWPUEPHG-UHFFFAOYSA-N
Synonym 1-4-trifluoromethyl-2-pyrimidinyl-4-piperidinecarboxylic acid, 1-4-trifluoromethyl pyrimidin-2-yl piperidine-4-carboxylic acid, 1-4-trifluoromethyl-2-pyrimidinyl-4-piperidine carboxylic acid, 1-4-trifluoromethyl pyrimid-2-yl piperidine-4-carboxylic acid, 1-4-trifluoromethyl-pyrimidin-2-yl-4-piperidinecarboxylic acid
PubChem CID 2777798
IUPAC Name 1-[4-(trifluoromethyl)pyrimidin-2-yl]piperidine-4-carboxylic acid
SMILES OC(=O)C1CCN(CC1)C1=NC=CC(=N1)C(F)(F)F

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 275.23
Percent Purity 97%
Chemical Name or Material 1-[4-(Trifluoromethyl)-2-pyrimidinyl]-4-piperidinecarboxylic acid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.