missing translation for 'onlineSavingsMsg'
Learn More

1-(4-Trifluoromethyl-2-pyrimidinyl)-1H-pyrazole-4-sulfonyl chloride, 95%

Product Code. 15498455
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15498455 250 mg 250mg
15488455 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15498455 Supplier Thermo Scientific Alfa Aesar Supplier No. H33424.MD
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 1215564-15-0 | C8H4ClF3N4O2S | 312.651 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1215564-15-0
Molecular Formula C8H4ClF3N4O2S
Molecular Weight (g/mol) 312.651
MDL Number MFCD11505058
InChI Key SADUDBJMXGGXDE-UHFFFAOYSA-N
Synonym 1-4-trifluoromethyl-2-pyrimidinyl-1h-pyrazole-4-sulfonyl chloride, 1-4-trifluoromethyl pyrimidin-2-yl-1h-pyrazole-4-sulfonyl chloride, 1-4-trifluoromethyl pyrimidin-2-yl-1h-pyrazole-4-sulphonyl chloride, 1-4-trifluoromethyl pyrimidin-2-yl pyrazole-4-sulfonyl chloride, chloro 1-4-trifluoromethyl pyrimidin-2-yl pyrazol-4-yl sulfone
PubChem CID 51063957
IUPAC Name 1-[4-(trifluoromethyl)pyrimidin-2-yl]pyrazole-4-sulfonyl chloride
SMILES C1=CN=C(N=C1C(F)(F)F)N2C=C(C=N2)S(=O)(=O)Cl

Specifications

Melting Point 97°C to 99°C
Quantity 250 mg
UN Number UN3261
Sensitivity Moisture sensitive
Formula Weight 312.66
Percent Purity 95%
Chemical Name or Material 1-(4-Trifluoromethyl-2-pyrimidinyl)-1H-pyrazole-4-sulfonyl chloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.