missing translation for 'onlineSavingsMsg'
Learn More

1-(4-tert-Butylphenyl)-4-chloro-1-butanone, TRC

Product Code. 30138136
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30138136 1 g 1g
30137999 5 g 5g
30005763 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30138136 Supplier Toronto Research Chemicals Supplier No. TRCB6909301G
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Identificatori chimici

CAS 43076-61-5
Molecular Formula C14H19ClO
Molecular Weight (g/mol) 238.75
Synonym 4-Chloro-1-[4-(1,1-dimethylethyl)phenyl]-1-butanone, 4’-tert-Butyl-4-chlorobutyrophenone, p-tert-Butyl-ω-chlorobutyrophenone, 4-Chloro-4’-tert-butylbutyrophenone
IUPAC Name 1-(4-tert-butylphenyl)-4-chloro-butan-1-one
SMILES CC(C)(C)c1ccc(cc1)C(=O)CCCCl

Specifica

Quantity 1 g
InChI Formula InChI=1S/C14H19ClO/c1-14(2,3)12-8-6-11(7-9-12)13(16)5-4-10-15/h6-9H,4-5,10H2,1-3H3
Formula Weight 238.1124
Chemical Name or Material 1-(4-tert-Butylphenyl)-4-chloro-1-butanone
Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.