missing translation for 'onlineSavingsMsg'
Learn More

1-(4-Pyridyl)piperazine, 97%

Product Code. 11328686
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11328686 1 g 1g
11338686 5 g 5g
11348686 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11328686 Supplier Thermo Scientific Alfa Aesar Supplier No. L19396.03
 Restricted Item
Estimated Shipment: 28-09-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 1008-91-9 | C9H13N3 | 163.224 g/mol

1-(4-Pyridyl)piperazine is an important raw material and intermediate used in organic synthesis, pharmaceuticals and agrochemicals. It is also used to produce 1-pyridin-4-yl-4-(4-vinyl-benzyl)-piperazine with 1-(chloromethyl)-4-vinylbenzene by heating.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-(4-Pyridyl)piperazine is an important raw material and intermediate used in organic synthesis, pharmaceuticals and agrochemicals. It is also used to produce 1-pyridin-4-yl-4-(4-vinyl-benzyl)-piperazine with 1-(chloromethyl)-4-vinylbenzene by heating.

Solubility
Soluble in water.

Notes
Keep container tightly closed in a dry and well-ventilated place. Containers which are opened must be carefully resealed and kept upright to prevent leakage. Store in cool place. Store under inert gas. It is sensitive to air. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1008-91-9
Molecular Formula C9H13N3
Molecular Weight (g/mol) 163.224
MDL Number MFCD00040745
InChI Key OQZBAQXTXNIPRA-UHFFFAOYSA-N
Synonym 1-4-pyridyl piperazine, 1-pyridin-4-yl piperazine, 1-pyridin-4-yl-piperazine, 4-piperazinopyridine, 4-pyridylpiperazine, 1-4-pyridyl-piperazine, 4-1-piperazino pyridine, 1-4-pyridinyl piperazine, piperazine, 1-4-pyridinyl
PubChem CID 70517
IUPAC Name 1-pyridin-4-ylpiperazine
SMILES C1CN(CCN1)C2=CC=NC=C2

Specifications

Melting Point 136°C to 141°C
Boiling Point 195°C to 200°C (0.3 mmHg)
Quantity 1 g
UN Number UN3259
Beilstein 609705
Formula Weight 163.22
Percent Purity 97%
Chemical Name or Material 1-(4-Pyridyl)piperazine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.