missing translation for 'onlineSavingsMsg'
Learn More

1,4-Phenylenediacryloyl chloride, tech., Thermo Scientific™

Product Code. 15457937
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15457937 250 mg 250mg
15437937 1 g 1g
15447937 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15457937 Supplier Thermo Scientific Alfa Aesar Supplier No. H51055.MD

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 35288-49-4
Molecular Formula C12H8Cl2O2
Molecular Weight (g/mol) 255.094
MDL Number MFCD01109676
InChI Key WQZLRZFBCIUDFL-KQQUZDAGSA-N
Synonym 1,4-phenylenediacryloyl chloride, 3,3'-1,4-phenylene bis 2-propenoyl chloride, 3,3'-benzene-1,4-diylbisprop-2-enoyl chloride, 1 4-phenylenediacryloyl chloridetech, 3,3'-p-phenylene bis acrylic acid chloride, 1,4-phenylenediacryloyl chloride, technical grade, 2-propenoyl chloride, 3,3'-1,4-phenylene bis, 3,3'-p-phenylene bis e-acrylic acid chloride, 2e,2'e-3,3'-1,4-phenylene diacryloyl chloride, e-3-4-e-3-chloro-3-oxoprop-1-enyl phenyl prop-2-enoyl chloride
PubChem CID 5989690
IUPAC Name (E)-3-[4-[(E)-3-chloro-3-oxoprop-1-enyl]phenyl]prop-2-enoyl chloride
SMILES C1=CC(=CC=C1C=CC(=O)Cl)C=CC(=O)Cl

Specifications

Quantity 250 mg
UN Number UN1759
Sensitivity Moisture sensitive
Formula Weight 255.1
Grade Technical
Chemical Name or Material 1,4-Phenylenediacryloyl chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.