missing translation for 'onlineSavingsMsg'
Learn More

1,4-Phenylene diisocyanide, 97%

Product Code. 15335088 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
1 g
Unit Size:
1g
This item is not returnable. View return policy

Product Code. 15335088

Brand: Thermo Scientific Acros 457080010

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 935-16-0 | C8H4N2 | 128.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 935-16-0
Melting Point 165.0°C to 166.0°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass Bottle
Molecular Formula C8H4N2
Quantity 1 g
Synonym 1,4-phenylene diisocyanide, 1,4-phenylenediisocyanide, tos-bb-0782, acmc-20ap1u, benzene,1,4-diisocyano, benzene, 1,4-diisocyano-9ci
InChI Key IXACFSRTSHAQIX-UHFFFAOYSA-N
SMILES [C-]#[N+]C1=CC=C(C=C1)[N+]#[C-]
IUPAC Name 1,4-diisocyanobenzene
Molecular Weight (g/mol) 128.13
PubChem CID 2771554
Formula Weight 128.13
Percent Purity 97%
Chemical Name or Material 1,4-Phenylene diisocyanide
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.