missing translation for 'onlineSavingsMsg'
Learn More

1,4-Pentadiene, 99%

Product Code. 10376960 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10376960 5 g Glass bottle 5g
10483395 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10376960 Supplier Thermo Scientific Acros Supplier No. 129860050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 591-93-5 | C5H8 | 68.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 591-93-5
Molecular Formula C5H8
Molecular Weight (g/mol) 68.11
MDL Number MFCD00008665
InChI Key QYZLKGVUSQXAMU-UHFFFAOYSA-N
Synonym 1,4-pentadiene, unii-a5if861oq5, pentadiene-1,4, allylethylene, penta-1,4-dien-1-yl, penta-1,4-dien-3-yl, ch2=chch2ch=ch2, acmc-1at58
PubChem CID 11587
IUPAC Name penta-1,4-diene
SMILES C=CCC=C

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point -148.0°C
Color Colorless
Density 0.6500g/mL
Boiling Point 26.0°C
Flash Point −20°C
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Linear Formula H2C=CHCH2CH=CH2
Refractive Index 1.3884 to 1.3904
Quantity 5 g
Beilstein 01, 251
Fieser 01,783
Specific Gravity 0.65
Formula Weight 68.11
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 1,4-Pentadiene, 99%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 1,4-Pentadiene
Signal Word
  • Danger
Hazard Category
  • Aspiration toxicity Category 1
  • Flammable liquid Category 1
Hazard Statement
  • H304-May be fatal if swallowed and enters airways.
  • H224-Extremely flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
  • P243-Take precautionary measures against static discharge.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P331-Do NOT induce vomiting.
  • P403+P235-Store in a well-ventilated place. Keep cool.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.