missing translation for 'onlineSavingsMsg'
Learn More

1,4-Oxazepan-5-one, 97%

Product Code. 11440527
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11440527 1 g 1g
11450527 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11440527 Supplier Thermo Scientific Alfa Aesar Supplier No. B22406.03
 Restricted Item
Estimated Shipment: 31-07-2026
Add to basket
This item is not returnable. View return policy

CAS: 10341-26-1 | C5H9NO2 | 115.13 g/mol

Used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Used as a pharmaceutical intermediate.

Notes
Hygroscopic. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10341-26-1
Molecular Formula C5H9NO2
Molecular Weight (g/mol) 115.13
MDL Number MFCD02683091
InChI Key OXDIYMHNQAWSCL-UHFFFAOYSA-N
Synonym tetrahydro-1,4-oxazepin-5 2h-one, 1,4-oxazepin-5 2h-one, tetrahydro, 4-oxacaprolactam, acmc-1bsly, tetrahydro-1,4-oxazepin-5 4h-one, tetrahydro-2h-1,4-oxazepin-5-one
PubChem CID 7023020
IUPAC Name 1,4-oxazepan-5-one
SMILES O=C1CCOCCN1

Specifications

Melting Point 80°C to 82°C
Quantity 1 g
Sensitivity Hygroscopic
Formula Weight 115.13
Percent Purity 97%
Chemical Name or Material 1,4-Oxazepan-5-one

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.