missing translation for 'onlineSavingsMsg'
Learn More

1-(4-Nitrophenylazo)-2-naphthol

Product Code. 11400458
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11400458 25 g 25g
11410458 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11400458 Supplier Thermo Scientific Alfa Aesar Supplier No. B24600.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 6410-10-2 | C16H11N3O3 | 293.28 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6410-10-2
Molecular Formula C16H11N3O3
Molecular Weight (g/mol) 293.28
MDL Number MFCD00003908
InChI Key ACHQDMGGRZPFAW-VLGSPTGOSA-N
ChEBI CHEBI:7986

Specifications

Melting Point 248°C to 252°C
Quantity 25 g
Beilstein 680469
Synonym pentoxifylline, trental, oxpentifylline, pentoxifyllin, torental, pentoxyphylline, pentoxil, dimethyloxohexylxanthine, pentoxiphyllium, vazofirin
SMILES [O-][N+](=O)C1=CC=C(N\N=C2/C(=O)C=CC3=CC=CC=C23)C=C1
IUPAC Name (1Z)-1-[2-(4-nitrophenyl)hydrazin-1-ylidene]-1,2-dihydronaphthalen-2-one
PubChem CID 4740
Formula Weight 293.28
Chemical Name or Material 1-(4-Nitrophenylazo)-2-naphthol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.