missing translation for 'onlineSavingsMsg'
Learn More

1-(4-Nitrophenyl)ethanol, TRC

Product Code. 30474924
Change view
Click to view available options
Quantity:
50 mg
500 mg
Unit Size:
500mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30474924 500 mg 500mg
30484178 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30474924 Supplier Toronto Research Chemicals Supplier No. TRCN545403500MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifikationer

Chemical Name or Material 1-(4-Nitrophenyl)ethanol
CAS 6531-13-1
Molecular Formula C8H9NO3
Quantity 500 mg
InChI Formula InChI=1S/C8H9NO3/c1-6(10)7-2-4-8(5-3-7)9(11)12/h2-6,10H,1H3
SMILES CC(O)c1ccc(cc1)[N+](=O)[O-]
IUPAC Name 1-(4-nitrophenyl)ethanol
Molecular Weight (g/mol) 167.16
Formula Weight 167.0582
Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.