missing translation for 'onlineSavingsMsg'
Learn More

1,4-Naphthoquinone, 97+% (dry wt.), cont. up to 5% water

Product Code. p-7081128
Click to view available options
Quantity:
100 g
500 g
2500 g
Unit Size:
100g
2500g
500g
This item is not returnable. View return policy

Product Code. 11454383

Brand: Thermo Scientific Alfa Aesar A10958.22

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 130-15-4 | C10H6O2 | 158.156 g/mol

1,4-Naphthoquinone is used as a strong dienophile in Diels-Alder reaction. It is also used as a precursor to anthroquinone and 5-nitro-1,4-naphthalenedione, which finds application in the preparation of aminoanthroquinone and is also used as a dye precursor. It is a basic component of Vitamin K.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,4-Naphthoquinone is used as a strong dienophile in Diels-Alder reaction. It is also used as a precursor to anthroquinone and 5-nitro-1,4-naphthalenedione, which finds application in the preparation of aminoanthroquinone and is also used as a dye precursor. It is a basic component of Vitamin K.

Solubility
Soluble in alkali hydroxide solvents, sparingly soluble in petroleum ether and water. Freely soluble in carbondioxide, acetic acid, benzene and choloroform.

Notes
Light sensitive. Incompatible with strong oxidizing agents and strong reducing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 130-15-4
Molecular Formula C10H6O2
Molecular Weight (g/mol) 158.156
MDL Number MFCD00001676
InChI Key FRASJONUBLZVQX-UHFFFAOYSA-N
Synonym 1,4-naphthoquinone, 1,4-naphthalenedione, p-naphthoquinone, naphthoquinone, alpha-naphthoquinone, 1,4-naphthylquinone, usaf cy-10, 1,4-dihydronaphthalene-1,4-dione, 1,4-dihydro-1,4-diketonaphthalene, 1,4-naftochinon
PubChem CID 8530
ChEBI CHEBI:27418
IUPAC Name naphthalene-1,4-dione
SMILES C1=CC=C2C(=O)C=CC(=O)C2=C1

Specifications

Melting Point 120°C to 124°C
Density 1.42
Flash Point 142°C (288°F)
Odor Irritating
Quantity 100 g
UN Number UN2811
Beilstein 878524
Merck Index 14,6395
Solubility Information Soluble in alkali hydroxide solvents,sparingly soluble in petroleum ether and water. Freely soluble in carbondioxide,acetic acid,benzene and choloroform.
Formula Weight 158.15
Percent Purity ≥97%
Concentration or Composition (by Analyte or Components) Contains up to 5% water
Assay (dry wt.)
Chemical Name or Material 1,4-Naphthoquinone
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.