missing translation for 'onlineSavingsMsg'
Learn More

1-(4-Methylphenyl)homopiperazine monohydrochloride, 98%, Thermo Scientific™

Product Code. 15499315
Change view
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15499315 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15499315 Supplier Thermo Scientific Alfa Aesar Supplier No. H51750.ME

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
1-(4-Methylphenyl)homopiperazine monohydrochloride is useful intermediate for synthesis.

Notes
Sold in collaboration with Tosoh Organic Chemical Co., Ltd.
TRUSTED_SUSTAINABILITY

Specifications

CAS 1192191-39-1
Melting Point 177°C to 181°C
Molecular Formula C12H19ClN2
MDL Number MFCD16172201
Quantity 500 mg
Beilstein 19720425
Synonym 1-4-methylphenyl homopiperazine monohydrochloride, 1-4-methylphenyl-1,4-diazepane hydrochloride, 4-mphp?hcl
InChI Key WOAJKNSSUFZJHK-UHFFFAOYSA-N
SMILES Cl.CC1=CC=C(C=C1)N1CCCNCC1
IUPAC Name 1-(4-methylphenyl)-1,4-diazepane;hydrochloride
Molecular Weight (g/mol) 226.75
PubChem CID 73012334
Formula Weight 226.75
Percent Purity 98%
Chemical Name or Material 1-(4-Methylphenyl)homopiperazine monohydrochloride
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.