missing translation for 'onlineSavingsMsg'
Learn More

1-(4-Methylphenyl)ethanol, 97%

Product Code. 11496390
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11496390 25 g 25g
11476390 1 g 1g
11486390 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11496390 Supplier Thermo Scientific Alfa Aesar Supplier No. B20832.14
 Restricted Item
Estimated Shipment: 03-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 536-50-5 | C9H12O | 136.194 g/mol

1-(4-Methylphenyl)ethanol serves as a component of the essential oil from some plants in the ginger family. It is also used as a flavoring agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-(4-Methylphenyl)ethanol serves as a component of the essential oil from some plants in the ginger family. It is also used as a flavoring agent.

Solubility
Slightly miscible with water.

Notes
Incompatible with oxidizing agents and strong reducing agents. Store in a cool place.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 536-50-5
Molecular Formula C9H12O
Molecular Weight (g/mol) 136.194
MDL Number MFCD00016860
InChI Key JESIHYIJKKUWIS-UHFFFAOYSA-N
Synonym 1-4-methylphenyl ethanol, 1-p-tolyl ethanol, bilagen, galinex, norbilan, tomobil, methyl-p-tolylcarbinol, p-tolylmethylcarbinol, 1-4-tolyl ethanol, 1-p-tolyl-1-ethanol
PubChem CID 10817
IUPAC Name 1-(4-methylphenyl)ethanol
SMILES CC1=CC=C(C=C1)C(C)O

Specifications

Density 0.987
Boiling Point 218°C to 220°C
Flash Point 107°C (225°F)
Refractive Index 1.523
Quantity 25 g
Beilstein 1857562
Merck Index 14,3237
Solubility Information Slightly miscible with water.
Formula Weight 136.2
Percent Purity 97%
Chemical Name or Material 1-(4-Methylphenyl)ethanol
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.