missing translation for 'onlineSavingsMsg'
Learn More

1-(4-Methyl-2-pyridyl)piperazine, Thermo Scientific™

Catalog Number 11499550
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
34803-67-3
C10H15N3
177.251
MFCD03412118
ZFBRKSGGMODDHD-UHFFFAOYSA-N
1-4-methylpyridin-2-yl piperazine, 1-4-methyl-pyridin-2-yl-piperazine, n-4-methylpyridin-2-yl-piperazine, 1-4-methyl-2-pyridyl piperazine, 1-4-methyl-2-pyridinyl piperazine, 1-2-4-methylpyridyl-piperazine, piperazine, 1-4-methyl-2-pyridinyl, 4-methyl-2-pyridyl piperazine, 1-2-4-methylpyridyl piperazine
2760058
1-(4-methylpyridin-2-yl)piperazine
CC1=CC(=NC=C1)N2CCNCC2
This item is not returnable. View return policy
34803-67-3
C10H15N3
177.251
MFCD03412118
ZFBRKSGGMODDHD-UHFFFAOYSA-N
1-4-methylpyridin-2-yl piperazine, 1-4-methyl-pyridin-2-yl-piperazine, n-4-methylpyridin-2-yl-piperazine, 1-4-methyl-2-pyridyl piperazine, 1-4-methyl-2-pyridinyl piperazine, 1-2-4-methylpyridyl-piperazine, piperazine, 1-4-methyl-2-pyridinyl, 4-methyl-2-pyridyl piperazine, 1-2-4-methylpyridyl piperazine
2760058
1-(4-methylpyridin-2-yl)piperazine
CC1=CC(=NC=C1)N2CCNCC2

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Quantity 250 mg
UN Number UN3259
Formula Weight 177.25
Chemical Name or Material 1-(4-Methyl-2-pyridyl)piperazine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'