missing translation for 'onlineSavingsMsg'
Learn More

1-(4-Methoxyphenyl)imidazole, 98%

Product Code. 11451090
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11451090 5 g 5g
11441090 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11451090 Supplier Thermo Scientific Alfa Aesar Supplier No. B25621.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 10040-95-6 | C10H10N2O | 174.203 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10040-95-6
Molecular Formula C10H10N2O
Molecular Weight (g/mol) 174.203
MDL Number MFCD00060492
InChI Key XNLOIFUGGCCEQX-UHFFFAOYSA-N
PubChem CID 82328
IUPAC Name 1-(4-methoxyphenyl)imidazole
SMILES COC1=CC=C(C=C1)N2C=CN=C2

Specifications

Melting Point 64°C to 67°C
Boiling Point 120°C (0.1 mmHg)
Odor Odorless
Quantity 5 g
Synonym 1-4-methoxyphenyl-1h-imidazole, 1-4-methoxyphenyl imidazole, n-4-methoxyphenyl imidazole, 1h-imidazole, 1-4-methoxyphenyl, 1-4-methoxy-phenyl-1h-imidazole, acmc-20ahwg, 4-imidazol-1-yl anisole, 4-imidazol-1-yl-anisole, 1-imidazolyl-4-methoxybenzene, 1h-imidazole,1-4-methoxyphenyl
Formula Weight 174.2
Percent Purity 98%
Chemical Name or Material 1-(4-Methoxyphenyl)imidazole

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.