missing translation for 'onlineSavingsMsg'
Learn More

1-(4-Methoxyphenyl)ethanol, 95%

Product Code. 11328595
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11328595 100 g 100g
11318595 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11328595 Supplier Thermo Scientific Alfa Aesar Supplier No. L12062.22
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 3319-15-1 | C9H12O2 | 152.19 g/mol

1-(4-Methoxyphenyl)ethanol is used to study the steady-state and nanosecond, laser-flash photolysis. It is used to produce 4-(1-chloro-ethyl)-anisole.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-(4-Methoxyphenyl)ethanol is used to study the steady-state and nanosecond, laser-flash photolysis. It is used to produce 4-(1-chloro-ethyl)-anisole.

Solubility
Not miscible in water.

Notes
Store in a cool, dry conditions in well sealed container. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3319-15-1
Molecular Formula C9H12O2
Molecular Weight (g/mol) 152.19
MDL Number MFCD00016857,MFCD09863666,MFCD09863667
InChI Key IUUULXXWNYKJSL-UHFFFAOYNA-N
Synonym 1-4-methoxyphenyl ethanol, 1-4-methoxyphenyl ethan-1-ol, 4-methoxy-alpha-methylbenzyl alcohol, 4-methoxyphenyl methyl carbinol, 1-p-methoxyphenyl ethanol, 4-methoxy-.alpha.-methylbenzyl alcohol, 1-4-methoxyphenyl-ethanol, acmc-20apae, 4-methoxyphenyl ethanol, 1 p-methoxyphenyl ethanol
PubChem CID 101148
ChEBI CHEBI:86566
IUPAC Name 1-(4-methoxyphenyl)ethanol
SMILES COC1=CC=C(C=C1)C(C)O

Specifications

Density 1.079
Boiling Point 94°C to 96°C (1 mmHg)
Flash Point 74°C (165°F)
Refractive Index 1.533
Quantity 100 g
Beilstein 2043521
Solubility Information Not miscible in water.
Formula Weight 152.19
Percent Purity 95%
Chemical Name or Material 1-(4-Methoxyphenyl)ethanol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.