missing translation for 'onlineSavingsMsg'
Learn More

1-(4-Methoxybenzyl)piperazine, 97%

Product Code. p-7052638
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
This item is not returnable. View return policy

Product Code. 15411248

Brand: Thermo Scientific Alfa Aesar H55099.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 21867-69-6 | C12H20N2O | 208.30 g/mol

1-(4-Methoxybenzyl)piperazine is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-(4-Methoxybenzyl)piperazine is used as a pharmaceutical intermediate.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 21867-69-6
Molecular Formula C12H20N2O
Molecular Weight (g/mol) 208.30
MDL Number MFCD01075236
InChI Key MGLUVVBFISROAH-UHFFFAOYSA-P
PubChem CID 231905
IUPAC Name 1-[(4-methoxyphenyl)methyl]piperazine
SMILES COC1=CC=C(C[NH+]2CC[NH2+]CC2)C=C1

Specifications

Melting Point 32°C to 36°C
Color White to Yellow
Flash Point >110°C (230°F)
Quantity 1 g
Synonym 1-4-methoxybenzyl piperazine, 1-4-methoxyphenyl methyl piperazine, piperazine,1-4-methoxyphenyl methyl, 1-4-methoxy-benzyl-piperazine, piperazine, 1-4-methoxyphenyl methyl, 4-methoxy-1-piperazinylmethyl benzene, 1-4-methoxybenzyl-piperazine, 1-4-methoxybenzyl piperazine #, piperazine, 1-4-methoxybenzyl, 1-4-methoxy-phenylmethyl piperazine
Formula Weight 206.29
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 1-(4-Methoxybenzyl)piperazine

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.