missing translation for 'onlineSavingsMsg'
Learn More

1-(4-Iodophenyl)-1H-pyrazole, ≥95%, Thermo Scientific™

Product Code. 10354422
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 368869-86-7
Molecular Formula C9H7IN2
Molecular Weight (g/mol) 270.07
MDL Number MFCD02681941
InChI Key ADIQBEMLKKKISJ-UHFFFAOYSA-N
PubChem CID 2776481
IUPAC Name 1-(4-iodophenyl)pyrazole
SMILES IC1=CC=C(C=C1)N1C=CC=N1

Specifications

CAS Min % 94.5
CAS Max % 100.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 1 g
Synonym 1-4-iodophenyl-1h-pyrazole, 1-4-iodophenyl pyrazole, 4-iodophenyl pyrazole, 1-4-iodo-phenyl-1h-pyrazole, 1h-pyrazole,1-4-iodophenyl, 1h-pyrazole, 1-4-iodophenyl
Formula Weight 270.07
Percent Purity 95%
Chemical Name or Material 1-(4-Iodophenyl)-1H-pyrazole
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'