missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-(4-Iodophenyl)-1H-pyrazole, ≥95%, Thermo Scientific™
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Specifications
CAS Min % | 94.5 |
CAS Max % | 100.0 |
Assay Percent Range | 95% |
Packaging | Amber Glass Bottle |
Quantity | 250 mg |
Synonym | 1-4-iodophenyl-1h-pyrazole, 1-4-iodophenyl pyrazole, 4-iodophenyl pyrazole, 1-4-iodo-phenyl-1h-pyrazole, 1h-pyrazole,1-4-iodophenyl, 1h-pyrazole, 1-4-iodophenyl |
Formula Weight | 270.07 |
Percent Purity | 95% |
Chemical Name or Material | 1-(4-Iodophenyl)-1H-pyrazole |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'