missing translation for 'onlineSavingsMsg'
Learn More

1-(4-Iodobenzyl)pyrrolidine, ≥97%, Thermo Scientific™

Product Code. 10200465
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10200465 5 g 5g
10642295 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10200465 Supplier Thermo Scientific Maybridge Supplier No. CC55611DE
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 858676-60-5
Molecular Formula C11H14IN
Molecular Weight (g/mol) 287.144
MDL Number MFCD07775752
InChI Key RDHZJYCRHTZBIX-UHFFFAOYSA-N
Synonym 1-4-iodobenzyl pyrrolidine, 1-4-iodophenyl methyl pyrrolidine, 1-4-iodo-benzyl-pyrrolidine, pyrrolidine,1-4-iodophenyl methyl
PubChem CID 18525867
IUPAC Name 1-[(4-iodophenyl)methyl]pyrrolidine
SMILES C1CCN(C1)CC2=CC=C(C=C2)I

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Quantity 5 g
Formula Weight 287.14
Percent Purity 97%
Chemical Name or Material 1-(4-Iodobenzyl)pyrrolidine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.