missing translation for 'onlineSavingsMsg'
Learn More

1-(4-Fluorophenyl)piperazine, 98%

Product Code. p-2694073 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
10 g
50 g
Packaging:
Glass bottle
Unit Size:
10g
50g
This item is not returnable. View return policy

Product Code. 10695252

Brand: Thermo Scientific Acros 184540100

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 2252-63-3 | C10H13FN2 | 180.22 g/mol

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2252-63-3
Molecular Formula C10H13FN2
Molecular Weight (g/mol) 180.22
InChI Key AVJKDKWRVSSJPK-UHFFFAOYSA-N
Synonym 1-4-fluorophenyl piperazine, 4-fluorophenylpiperazine, pfpp, 4-fluoro-phenylpiperazine, n-4-fluorophenyl piperazine, para-fluorophenylpiperazine, unii-vml61be244, 1-p-fluorophenyl piperazine, piperazine, 1-4-fluorophenyl, 4-fluorophenyl piperazine
PubChem CID 75260
IUPAC Name 1-(4-fluorophenyl)piperazine
SMILES C1CN(CCN1)C2=CC=C(C=C2)F

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color Yellow
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.5585 to 1.5605
Quantity 10 g
Formula Weight 180.22
Percent Purity 98%
Physical Form Liquid After Melting
Chemical Name or Material 1-(4-Fluorophenyl)piperazine
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 1-(4-Fluorophenyl)piperazine
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 C
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H312-Harmful in contact with skin.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P264-Wash thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.