missing translation for 'onlineSavingsMsg'
Learn More

1-(4-Fluorophenyl)ethanol, 98%

Product Code. 11499597
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11499597 100 g 100g
11479597 5 g 5g
11489597 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11499597 Supplier Thermo Scientific Alfa Aesar Supplier No. B22097.22

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 403-41-8 | C8H9FO | 140.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 403-41-8
Molecular Formula C8H9FO
Molecular Weight (g/mol) 140.16
MDL Number MFCD00004515,MFCD03092997,MFCD03092998
InChI Key PSDSORRYQPTKSV-UHFFFAOYNA-N
Synonym 1-4-fluorophenyl ethanol, 4-fluoro-alpha-methylbenzyl alcohol, 1-4-fluorophenyl ethan-1-ol, 1-p-fluorophenyl ethanol, p-fluorophenylmethylcarbinol, 4-fluorophenylmethylcarbinol, 1-4-fluorophenyl-1-ethanol, alpha-methyl-4-fluorobenzyl alcohol, 1-4-fluorophenyl-ethanol, 4-fluorophenyl methyl carbinol
PubChem CID 73946
IUPAC Name 1-(4-fluorophenyl)ethanol
SMILES CC(O)C1=CC=C(F)C=C1

Specifications

Density 1.109
Boiling Point 90°C to 92°C (7 mmHg)
Flash Point 91°C (196°F)
Quantity 100 g
Beilstein 2042025
Formula Weight 140.16
Percent Purity 98%
Chemical Name or Material 1-(4-Fluorophenyl)ethanol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.