missing translation for 'onlineSavingsMsg'
Learn More

1-(4-Fluorophenyl)-5-methyl-1H-pyrazole-4-carbaldehyde, 97%, Thermo Scientific™

Product Code. 10160692
Change view
Click to view available options
Quantity:
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10160692 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10160692 Supplier Thermo Scientific Maybridge Supplier No. MO00310DA
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Specifications

CAS 423768-41-6
CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Molecular Formula C11H9FN2O
MDL Number MFCD03407349
Quantity 1 g
Synonym 1-4-fluorophenyl-5-methyl-1h-pyrazole-4-carbaldehyde, 1-4-fluorophenyl-5-methyl-1h-pyrazole-4-carboxaldehyde, 1-4-fluorophenyl-5-methylpyrazole-4-carbaldehyde, acmc-20apar
InChI Key XFVNWAPZODECJD-UHFFFAOYSA-N
SMILES CC1=C(C=NN1C2=CC=C(C=C2)F)C=O
IUPAC Name 1-(4-fluorophenyl)-5-methylpyrazole-4-carbaldehyde
Molecular Weight (g/mol) 204.204
PubChem CID 2780707
Formula Weight 204.2
Percent Purity 97%
Chemical Name or Material 1-(4-Fluorophenyl)-5-methyl-1H-pyrazole-4-carbaldehyde
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.