missing translation for 'onlineSavingsMsg'
Learn More

[1-(4-Fluorophenyl)-5-methyl-1H-pyrazol-4-yl]methanol, 97%, Thermo Scientific™

Product Code. p-2693845
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10354242 250 mg Amber Glass Bottle 250mg
10752873 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10354242 Supplier Thermo Scientific Maybridge Supplier No. MO00765CB
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 465514-37-8
Molecular Formula C11H11FN2O
Molecular Weight (g/mol) 206.22
MDL Number MFCD03644148
InChI Key QJPGMVFNIWHOIY-UHFFFAOYSA-N
Synonym 1-4-fluorophenyl-5-methyl-1h-pyrazol-4-yl methanol, 1-4-fluorophenyl-5-methylpyrazol-4-yl methanol, fmq, maybridge3_004281, 1-4-fluorophenyl-5-methyl-4-pyrazolyl methanol, 1-4-fluorophenyl-5-methyl-pyrazol-4-yl methanol, 1-4-fluorophenyl-5-methyl-1h-pyrazole-4-methanol, 1-4-fluorophenyl-5-methylpyrazol-4-yl methan-1-ol
PubChem CID 2780717
IUPAC Name [1-(4-fluorophenyl)-5-methylpyrazol-4-yl]methanol
SMILES CC1=C(CO)C=NN1C1=CC=C(F)C=C1

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 206.22
Percent Purity 97%
Chemical Name or Material [1-(4-Fluorophenyl)-5-methyl-1H-pyrazol-4-yl]methanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.