missing translation for 'onlineSavingsMsg'
Learn More

1-(4-Fluorophenyl)-1,3-dihydro-3-oxo-5-isobenzofurancarboxamide, TRC

Product Code. 30390070
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30390070 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30390070 Supplier Toronto Research Chemicals Supplier No. TRCF59529010MG
 Restricted Item
Estimated Shipment: 25-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

CAS 372941-46-3
Molecular Formula C15 H10 F N O3
Quantity 10 mg
Synonym 5-Isobenzofurancarboxamide, 1-(4-fluorophenyl)-1,3-dihydro-3-oxo-, 1-(4-Fluorophenyl)-1,3-dihydro-3-oxo-5-isobenzofurancarboxamide, 1-(4-Fluorophenyl)-3-oxo-1,3-dihydro-2-benzofuran-5-carboxamide, 1-(4-Fluorophenyl)-3-oxo-1,3-dihydroisobenzofuran-5-carboxamide
InChI Formula InChI=1S/C15H10FNO3/c16-10-4-1-8(2-5-10)13-11-6-3-9(14(17)18)7-12(11)15(19)20-13/h1-7,13H,(H2,17,18)
SMILES NC(=O)c1ccc2C(OC(=O)c2c1)c3ccc(F)cc3
IUPAC Name 1-(4-fluorophenyl)-3-oxo-1H-2-benzofuran-5-carboxamide
Molecular Weight (g/mol) 271.24
Formula Weight 271.0645
Chemical Name or Material 1-(4-Fluorophenyl)-1,3-dihydro-3-oxo-5-isobenzofurancarboxam
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.