Learn More
1-(4-Fluorobenzyl)-5-oxo-3-pyrrolidinecarboxylic acid, 97%, Maybridge™
Brand: Maybridge MO00127DA
Code : 31
Additional Details : CAS Number : 96449-68-2 Weight : 0.05100kg Transport : UN number : 2811 Chem class : 6.1 Pack group : III
Quantity | 1g |
---|
Chemical Identifiers
96449-68-2 | |
237.23 | |
IYAFJFDPMVQTJO-UHFFFAOYSA-N | |
2780702 | |
C1C(CN(C1=O)CC2=CC=C(C=C2)F)C(=O)O |
C12H12FNO3 | |
MFCD03086215 | |
1-4-fluorobenzyl-5-oxopyrrolidine-3-carboxylic acid, 1-4-fluorophenyl methyl-5-oxopyrrolidine-3-carboxylic acid, 1-4-fluorobenzyl-5-oxo-3-pyrrolidinecarboxylic acid, 3-pyrrolidinecarboxylicacid, 1-4-fluorophenyl methyl-5-oxo, 1-4-fluorobenzyl-5-oxopyrrolidin-3-carboxylic acid, 1-4-fluorobenzyl-5-oxo-pyrrolidine-3-carboxylic acid, 1-4-fluorophenyl methyl-5-oxo-3-pyrrolidinecarboxylic acid, 1-4-fluorophenyl methyl-5-oxo-pyrrolidine-3-carboxylic acid, 1-4-fluorophenyl methyl-5-oxidanylidene-pyrrolidine-3-carboxylic acid | |
1-[(4-fluorophenyl)methyl]-5-oxopyrrolidine-3-carboxylic acid |
Specifications
1-(4-Fluorobenzyl)-5-oxo-3-pyrrolidinecarboxylic acid | |
96449-68-2 | |
97% | |
MFCD03086215 | |
IYAFJFDPMVQTJO-UHFFFAOYSA-N | |
1-[(4-fluorophenyl)methyl]-5-oxopyrrolidine-3-carboxylic acid | |
2780702 | |
97% |
1g | |
97.0 | |
C12H12FNO3 | |
1-4-fluorobenzyl-5-oxopyrrolidine-3-carboxylic acid, 1-4-fluorophenyl methyl-5-oxopyrrolidine-3-carboxylic acid, 1-4-fluorobenzyl-5-oxo-3-pyrrolidinecarboxylic acid, 3-pyrrolidinecarboxylicacid, 1-4-fluorophenyl methyl-5-oxo, 1-4-fluorobenzyl-5-oxopyrrolidin-3-carboxylic acid, 1-4-fluorobenzyl-5-oxo-pyrrolidine-3-carboxylic acid, 1-4-fluorophenyl methyl-5-oxo-3-pyrrolidinecarboxylic acid, 1-4-fluorophenyl methyl-5-oxo-pyrrolidine-3-carboxylic acid, 1-4-fluorophenyl methyl-5-oxidanylidene-pyrrolidine-3-carboxylic acid | |
C1C(CN(C1=O)CC2=CC=C(C=C2)F)C(=O)O | |
237.23 | |
237.23 |