missing translation for 'onlineSavingsMsg'
Learn More

1-(4-Fluoro-2-nitrophenyl)-4-piperidone, 97%, Thermo Scientific™

Product Code. 15470829
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1019582-23-0

Specifications

Quantity 250 mg
Formula Weight 238.22
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 1-(4-Fluoro-2-nitrophenyl)-4-piperidone
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'