missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-[(4-Ethoxyphenyl)ethynyl]-4-n-pentylbenzene, 99+%, Thermo Scientific™
£23.20 - £140.00
Chemical Identifiers
CAS | 95480-29-8 |
---|---|
Molecular Formula | C21H24O |
Molecular Weight (g/mol) | 292.422 |
MDL Number | MFCD01218020 |
InChI Key | QRMMWBMCJKXXKV-UHFFFAOYSA-N |
Synonym | 1-4-ethoxyphenyl ethynyl-4-n-pentylbenzene, 1-2-4-ethoxyphenyl ethynyl-4-pentylbenzene, 1-ethoxy-4-4-pentylphenyl ethynyl benzene, 1-ethoxy-4-2-4-pentylphenyl ethynyl benzene, 4-ethoxy-1-2-4-pentylphenyl ethynyl benzene, 1-4-ethoxyphenyl ethynyl-4-pentylbenzene #, benzene, 1-ethoxy-4-4-pentylphenyl ethynyl |
PubChem CID | 610223 |
IUPAC Name | 1-ethoxy-4-[2-(4-pentylphenyl)ethynyl]benzene |
SMILES | CCCCCC1=CC=C(C=C1)C#CC2=CC=C(C=C2)OCC |
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
95480-29-8 | |
292.422 | |
QRMMWBMCJKXXKV-UHFFFAOYSA-N | |
610223 | |
CCCCCC1=CC=C(C=C1)C#CC2=CC=C(C=C2)OCC |
C21H24O | |
MFCD01218020 | |
1-4-ethoxyphenyl ethynyl-4-n-pentylbenzene, 1-2-4-ethoxyphenyl ethynyl-4-pentylbenzene, 1-ethoxy-4-4-pentylphenyl ethynyl benzene, 1-ethoxy-4-2-4-pentylphenyl ethynyl benzene, 4-ethoxy-1-2-4-pentylphenyl ethynyl benzene, 1-4-ethoxyphenyl ethynyl-4-pentylbenzene #, benzene, 1-ethoxy-4-4-pentylphenyl ethynyl | |
1-ethoxy-4-[2-(4-pentylphenyl)ethynyl]benzene |
Specifications
95480-29-8 | |
178°C to 180°C (0.5 mmHg) | |
MFCD01218020 | |
QRMMWBMCJKXXKV-UHFFFAOYSA-N | |
1-ethoxy-4-[2-(4-pentylphenyl)ethynyl]benzene | |
610223 | |
≥99% |
62°C (N) | |
C21H24O | |
1-4-ethoxyphenyl ethynyl-4-n-pentylbenzene, 1-2-4-ethoxyphenyl ethynyl-4-pentylbenzene, 1-ethoxy-4-4-pentylphenyl ethynyl benzene, 1-ethoxy-4-2-4-pentylphenyl ethynyl benzene, 4-ethoxy-1-2-4-pentylphenyl ethynyl benzene, 1-4-ethoxyphenyl ethynyl-4-pentylbenzene #, benzene, 1-ethoxy-4-4-pentylphenyl ethynyl | |
CCCCCC1=CC=C(C=C1)C#CC2=CC=C(C=C2)OCC | |
292.422 | |
292.42 | |
1-[(4-Ethoxyphenyl)ethynyl]-4-n-pentylbenzene |
Safety and Handling
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures