missing translation for 'onlineSavingsMsg'
Learn More

1,4-Dithioerythritol, Electrophoresis Grade, 99%

Product Code. 15406509
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15406509 1 g 1g
15416509 5 g 5g
15426509 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15406509 Supplier Thermo Scientific Alfa Aesar Supplier No. J64656.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 6892-68-8 | C4H10O2S2 | 154.24 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6892-68-8
Molecular Formula C4H10O2S2
Molecular Weight (g/mol) 154.24
MDL Number MFCD00063750
InChI Key VHJLVAABSRFDPM-ZXZARUISSA-N
Synonym dithioerythritol, 1,4-dithioerythritol, dte, 2r,3s-1,4-dimercaptobutane-2,3-diol, erythro-1,4-dimercapto-2,3-butanediol, 2,3-butanediol, 1,4-dimercapto-, 2r,3s-rel, 1,4-dithioerythritol dte, 2r*,3s*-1,4-dimercapto-2,3-butanediol, erythro-2,3-dihydroxy-1,4-butanedithiol, 2r,3s-1,4-disulfanylbutane-2,3-diol
PubChem CID 439352
ChEBI CHEBI:17456
SMILES O[C@@H](CS)[C@H](O)CS

Specifications

Melting Point 82°C to 84°C
Color White
Odor Stench
Quantity 1 g
UN Number UN3335
Beilstein 1719756
Sensitivity Air sensitive
IUPAC Name (2R,3S)-1,4-bis(sulfanyl)butane-2,3-diol
Formula Weight 154.25
Percent Purity 99%
Grade Electrophoresis
Physical Form Powder
Chemical Name or Material 1,4-Dithioerythritol
Show More Show Less
compliance-icons
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.