missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methylated Spirit (IDA 99), 99% (v/v), Pure, (Industrial Methylated Spirit, 74 0.P.)
Shop All Fisher Chemical ProductsSpecifications
Specifications
| Melting Point | -114.1°C |
| Color | Colorless |
| Boiling Point | 78.2°C |
| Packaging | Amber glass bottle |
| Quantity | 2.5 L |
| Physical Form | Liquid |
Safety and Handling
Product Identifier
- Methylated spirit industrial
- Danger
- Flammable liquid Category 2
- Specific target organ toxicity Category 2
- Acute toxicity Category 4
- H225-Highly flammable liquid and vapour.
- H302-Harmful if swallowed.
- H312-Harmful in contact with skin.
- H319-Causes serious eye irritation.
- H332-Harmful if inhaled.
- H371-May cause damage to organs.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P240-Ground/bond container and receiving equipment.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- MIXTURE LIST-Contain: Ethyl alcohol, Methyl alcohol
Título del producto
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.
¿Detecta una oportunidad de mejora?