missing translation for 'onlineSavingsMsg'
Learn More

1,4-Dioxaspiro[4.5]decan-8-ol, 98%

Product Code. 10287543 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 10287543

Brand: Thermo Scientific Acros 241010050

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 22428-87-1 | C8H14O3 | 158.20 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 22428-87-1
Molecular Formula C8H14O3
Molecular Weight (g/mol) 158.20
MDL Number MFCD00067003
InChI Key HKQTYQDNCKMNHZ-UHFFFAOYSA-N
Synonym 1,4-dioxaspiro 4.5 decan-8-ol, 1,4-dioxa-spiro 4.5 decan-8-ol, 4-hydroxycyclohexanone ethylene acetal, 4-hydroxycyclohexanone monoethylene ketal, 8-hydroxy-1,4-dioxaspiro 4.5 decane, pubchem20110, acmc-1chv5, 4,4-ethylenedioxycyclohexanol
PubChem CID 567352
IUPAC Name 1,4-dioxaspiro[4.5]decan-8-ol
SMILES OC1CCC2(CC1)OCCO2

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color Colorless
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.4850 to 1.487
Quantity 5 g
Formula Weight 158.2
Percent Purity 98%
Physical Form Viscous Liquid
Chemical Name or Material 1, 4-Dioxaspiro[4.5]decan-8-ol
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.