missing translation for 'onlineSavingsMsg'
Learn More

1,4-Dioxane, Reag. Ph. Eur. reag. ISO, Solstice

Product Code. 15663910
Change view
Click to view available options
Quantity:
1 L
2.5 L
210 kg
Unit Size:
1 L
2.5 L
210kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15663910 210 kg 210kg
15643910 1 L 1 L
15653910 2.5 L 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15663910 Supplier Fluka Supplier No. 33147210KG
 Restricted Item
Request Bulk or Custom Formats
Currently Unavailable: We have no information when this product will be available.
View alternative products
This item is not returnable. View return policy

Chemical Identifiers

CAS 123-91-1
Molecular Formula C4H8O2
Molecular Weight (g/mol) 88.106
MDL Number MFCD00006571
InChI Key RYHBNJHYFVUHQT-UHFFFAOYSA-N
Synonym dioxane, p-dioxane, 1,4-diethylene dioxide, diethylene ether, dioxan, 1,4-dioxacyclohexane, diethylene dioxide, dioxanne, di ethylene oxide, tetrahydro-p-dioxin
PubChem CID 31275
ChEBI CHEBI:47032
IUPAC Name 1,4-dioxane
SMILES C1COCCO1

Specifications

Density 1.034g/mL at 25°C
Quantity 210 kg
Explosion Limit 0.22
Flash Point 12°C (53.6°F)
Ignition Point 356°F
Vapor Pressure 40mmHg (25°C), 27mmHg (20°C)
UN Number 1165
Beilstein 102551
Formula Weight 88.11g/mol
Percent Purity ≥99.5% (GC)
Grade ACS/puriss. p.a.
Chemical Name or Material 1,4-Dioxane, Reag. Ph. Eur. reag. ISO
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.