missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,4-Dioxane-d8, SPEX CertiPrep™
Click to view available options
Quantity:
1 mL
Packaging:
Amber Prescored Glass Ampule
Description
Includes
Autosampler Vial, NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet
Organic Standard manufactured for GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation
Analyte | CAS No. | Concentration |
---|---|---|
1,4-Dioxane-d8 | 17647-74-4 | 1000μg/mL |
Certifications
Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001
Compliance
GHS/OSHA Complaint
Specifications
Packaging | Amber Prescored Glass Ampule |
Quantity | 1 mL |
CAS | 17647-74-4, 67-56-1 |
CAS Min % | 0.1, 99.9 |
CAS Max % | 0.1, 99.9 |
Molecular Formula | C4H8O2 |
Synonym | 1,4-dioxane-d8, 2h8-1,4-dioxane, 1,4-dioxane-d8, 99 atom % d, 2 h?-1,4-dioxane, 1,4-dioxane-2lh8, 1,4-dioxane d8, 1,4-dioxane-d8, ≥99 atom % d, 1,4-dioxane-d isotopic 1g, 1,4-dioxane-d8 >99.0 atom % d, 1 4-dioxane-d8 99 atom % d |
InChI Key | RYHBNJHYFVUHQT-SVYQBANQSA-N |
SMILES | C1COCCO1 |
IUPAC Name | 2,2,3,3,5,5,6,6-octadeuterio-1,4-dioxane |
Show More |
Safety and Handling
Product Identifier
- 1,4-Dioxane-d8
- Danger
- Acute toxicity Category 3
- Flammable liquid Category 2
- Specific target organ toxicity after single exposure Category 1
- H225-Highly flammable liquid and vapour.
- H331-Toxic if inhaled.
- H370-Causes damage to organs.
- P101-If medical advice is needed, have product container or label at hand.
- P102-Keep out of reach of children.
- P103-Read label before use.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
- MIXTURE LIST-Contains : methanol
Product intended for laboratory use only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'